Learn More
Brady™ and TLS-PC Link™ Labels
Manufacturer:  Brady™ 18424
 View more versions of this product
Catalog No. 19-150-064
  Check Availability
Add to cart
Specifications
Description
Compliance
UL, CSA & AGA approved with its respective ribbon.
Description & Specifications
Specifications
Acrylic,Permanent
AGA Approved, CSA Approved, Halogen-free (DIN VDE 0472/parts 815), UL Recognized
0.75 in.
BMP™71, TLS 2200™, TLS-PC Link™
Label
250/Roll
Green,White
Gloss
Asset tracking, barcoding, component ID, equipment ID, general purpose ID, rating plates
1.9cm
Polyester