Learn More
Brady™ Accident Prevention Tags - Caution Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Brady™ 86712
 View more versions of this product
Catalog No. 19-040-843
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM D1593, OSHA 1910.145
0.37 in.
For temporary tagging in a non harsh environment
17.78cm
-40° to 194°F
0.25mm
10.16cm
Front: Do Not Operate Electricians At Work, Back: Do Not Remove This Tag
Black on Yellow
0.93cm
7 in.
100 string fasteners
-40° to 90°C
4 in.
Cardstock