Learn More
Brady™ Accident Prevention Tags - Danger Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Brady™ 86468
 View more versions of this product
Catalog No. 19-040-243
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
OSHA
0.37 in.
Up to 2 Years
3.9 Gardner
17.7cm
-40° to 175°F
0.02 to 0.025 in.
10.1cm
Danger Do Not Remove This Tag Pellagra No Sacra Learjet De Advertencies
Black/Red on White
0.93cm
For lockout and tagout identification, polyester laminate protects the graphics, wipes clean, and withstands water, grease, and extreme temperatures, heavy duty construction prevents accidental removal
7 in.
10 nylon ties
-40° to 80°C
4 in.
Heavy-duty Polyester