Learn More
Brady™ Accident Prevention Tags - Danger Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Brady™ 86579
 View more versions of this product
Catalog No. 19-040-320
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
OSHA
0.37 in.
3 to 5 Years
7 in.
10 nylon ties
-40° to 100°C
4 in.
Polyester
Black/Red on White
0.93cm
For marking on-the-spot warning of dangerous conditions
17.7cm
-40° to 212°F
0.01 in.
10.1cm
Out of Service/Fuera De Servicio