Learn More
Brady™ Accident Prevention Tags with Same Fronts and Backs
Feature same message on tag front and back
Supplier:  Brady™ 86478
 View more versions of this product
Catalog No. 19-040-257
Add to Cart
Description
Description
  • Feature two chamfered corners and a 3/8 in. grommeted hole
  • Textured write-on surface allows for authorized signatures, dates, and other special instructions
  • Chemical-resistant, plastic Dura-Tags are reusable and designed for harsh plant environments; come with nylon fasteners
  • Chemical-resistant economy polyester tags withstand moisture, grease and dirt; come with nylon fasteners
  • Economical, yet durable cardstock tags come with string fasteners
Compliance
Meet OSHA 1910.145 for accident prevention; Dura-Tag and polyester tags also meet OSHA 50 lb. pull requirement.
Specifications
Specifications
OSHA
Dura-Tag
For lockout and tagout identification, polyester laminate protects the graphics, wipes clean, and withstands water, grease, and extreme temperatures, heavy duty construction prevents accidental removal
3.9 Gardner
17.7cm
0.02 to 0.025 in.
-40°F to 175°F
10.1 cm
0.37 in.
Notice Temporary Out Of Use
Up to 2 Years
7 in.
10 nylon ties
-40°C to 80°C
4 in.
0.93 cm
Product Suggestions
Product Suggestions