Learn More
Brady™ Accident Prevention Tags with Same Fronts and Backs Available on GSA/VA Contract for Federal Government customers only.
Feature same message on tag front and back
Manufacturer:  Brady™ 86565
 View more versions of this product
Catalog No. 19-040-816
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
OSHA
0.93cm
For marking on-the-spot warning of dangerous conditions
14.6cm
-40° to 212°F
0.01 in.
7.6cm
Inspected Record Date/By/Accepted/Rejected
0.37 in.
3 to 5 Years
5.75 in.
10 nylon fasteners
-40° to 100°C
3 in.
Polyethylene