Learn More
Brady™ Accident Prevention Tags with Same Fronts and Backs Available on GSA/VA Contract for Federal Government customers only.
Feature same message on tag front and back
Manufacturer:  Brady™ 86438
 View more versions of this product
Catalog No. 19-040-389
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Dura-Tag
OSHA
0.93cm
For lockout and tagout identification, polyester laminate protects the graphics, wipes clean, and withstands water, grease, and extreme temperatures, heavy duty construction prevents accidental removal
5.75 in.
10 nylon fasteners
-40° to 80°C
3 in.
Inspected Record Date/By/Accepted/Rejected
0.37 in.
Up to 2 Years
3.9 Gardner
14.6cm
-40° to 175°F
0.02 to 0.025 in.
7.6cm