Learn More
Brady™ Accident Prevention Tags with Same Fronts and Backs
Feature same message on tag front and back
Supplier:  Brady™ 86556
 View more versions of this product
Catalog No. 19-040-427
Add to Cart
Description
Description
  • Feature two chamfered corners and a 3/8 in. grommeted hole
  • Textured write-on surface allows for authorized signatures, dates, and other special instructions
  • Chemical-resistant, plastic Dura-Tags are reusable and designed for harsh plant environments; come with nylon fasteners
  • Chemical-resistant economy polyester tags withstand moisture, grease and dirt; come with nylon fasteners
  • Economical, yet durable cardstock tags come with string fasteners
Compliance
Meet OSHA 1910.145 for accident prevention; Dura-Tag and polyester tags also meet OSHA 50 lb. pull requirement.
Specifications
Specifications
OSHA
Polyester
For marking on-the-spot warning of dangerous conditions
5.75 in.
10 nylon fasteners
-40°C to 100°C
3 in.
0.93 cm
0.37 in.
Safety Inspection Equipment ID/Location/Date/By
3 to 5 Years
14.6cm
0.01 in.
-40°F to 212°F
7.6 cm
Product Suggestions
Product Suggestions