Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Admittance Sign: DANGER - HIGH VOLTAGE KEEP OUT Available on GSA/VA Contract for Federal Government customers only.

Identify electrical hazard and high voltage areas

Manufacturer:  Brady™ 84084

 View more versions of this product

Catalog No. 19-103-651


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Material: Self-sticking Polyester
25.40cm
Admittance Sign
35.56cm
Polyester
10 in.
DANGER - HIGH VOLTAGE KEEP OUT
14 in.
Black/Red on White
1/Ea.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit