Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Admittance Sign: DANGER - HIGH VOLTAGE KEEP OUT

Identify electrical hazard and high voltage areas

$9.19 - $68.30

Specifications

Color Red on White
Product Type Admittance Sign
View More Specs

Products 4
Catalog Number Mfr. No. Material Width (English) Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Material Width (English) Height (English) Price Quantity & Availability  
19-041-629
View Documents
Brady™
22104
Plastic 14 in. 10 in.
Each for $31.25
Add to Cart
 
19-103-651
View Documents
Brady™
84084
Polyester 14 in. 10 in.
Each for $68.30
Add to Cart
 
19-000-657
View Documents
Brady™
40668
Aluminum 14 in. 10 in.
Each for $68.30
Add to Cart
 
19-835-116
View Documents
Brady™
84082
Polyester 3.5 in. 5 in.
Each for $9.19
Add to Cart
 
Description

Description

  • Available in two materials - plastic and self sticking polyester
  • Black/red on white

Specifications

Specifications

Red on White
Admittance Sign
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit