Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Admittance Signs: DANGER - ELECTRICAL EQUIPMENT - AUTHORIZED PERSONNEL - ONLY
Identify electrical hazard and high voltage areas
Supplier: Brady™ 71543
View more versions of this product
Safety Services Available
Overview
Logo/Customization
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in different widths and heights
- Black/red on white
Specifications
ANSI Z535, OSHA 1910.145, PSTC-33, | |
DANGER Electrical Equipment Authorized Personnel Only | |
-40°F to 193°F | |
1 Ea. | |
Surface Mount | |
7 in. | |
193°F | |
0.1 in. | |
25.40 cm |
Red on White | |
Admittance Sign | |
Fiberglass | |
9000 psi | |
Industrial, utility, commercial and institutional environments for both indoor and outdoor use | |
17.78 cm | |
-40°F | |
10 in. | |
-40°C to 90°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction