Brady™ Admittance Signs: DANGER - ELECTRICAL EQUIPMENT - AUTHORIZED PERSONNEL - ONLY Available on GSA/VA Contract for Federal Government customers only.

Identify electrical hazard and high voltage areas

Manufacturer: Brady™ 84069

 View more versions of this product

Catalog No. 19-035-595

  Check Availability
Add to Cart

    Specifications

    Height (English) 10 in.,10 in.
    Height (Metric) 25.40cm,25.40cm
    Material Polyester
    Quantity Each
    Style Surface Mount
    Temperature Range (Metric) -40 to 110°C
    Thickness 0.006 to 0.01 in.
    Width (English) 14 in.
    For Use With (Application) Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
    Temperature Range (English) -40 to 230°F
    Width (Metric) 35.56cm
    Certifications/Compliance ISO 9001, PSTC-33,ISO 9001, PSTC-33
    Message Danger Electrical Equipment Authorized Personnel Only
    Color Black, Red on White,Black, Red on White
    Type Admittance sign

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit