Brady™ Admittance Signs: DANGER - HIGH VOLTAGE Available on GSA/VA Contract for Federal Government customers only.

Identifies electrical hazard and high voltage areas

Manufacturer: Brady™ 84877

 View more versions of this product

Catalog No. 19-015816

  Check Availability
Add to Cart

    Specifications

    Height (English) 10 in.,10 in.
    Height (Metric) 25.4cm,25.4cm
    Material Self-sticking Polyester
    Certifications/Compliance ISO 9001, ANSI Z535, OSHA 1910.145, PSTC-33,,ISO 9001, ANSI Z535, OSHA 1910.145, PSTC-33,
    Message Danger High Voltage
    Quantity Each
    Style Surface Mount
    Temperature Range (English) -40° to +230°F
    Temperature Range (Metric) -40° to +110°C
    Thickness 0.006 to 0.01 in.
    Color Black, Red on White,Black, Red on White
    Type Admittance sign
    Width (English) 14 in.
    Width (Metric) 35.56cm
    For Use With (Application) Industrial, utility, commercial and institutional environments for both indoor and outdoor use

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit