Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Admittance Signs: DANGER - HIGH VOLTAGE
Identifies electrical hazard and high voltage areas
Supplier: Brady™ 84876
View more versions of this product
Safety Services Available
Overview
Logo/Customization
Description
- Uses bright colors, bold text and intuitive pictograms to warn against entry, machine operation and buried cables
- Available in three materials - fiberglass, plastic and self-sticking polyester
- Available in different widths and heights
- Black/red on white
Specifications
ISO 9001, ANSI Z535, OSHA 1910.145, PSTC-33, | |
1 Ea. | |
Surface Mount | |
7 in. | |
230°F | |
0.006 to 0.01 in. | |
25.4 cm | |
-40°F to 230°F | |
Admittance Sign |
Polyester | |
Self-sticking Polyester | |
Industrial, utility, commercial and institutional environments for both indoor and outdoor use | |
17.78 cm | |
-40°F | |
10 in. | |
-40°C to 110°C | |
DANGER High Voltage | |
Red on White |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction