Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Admittance Signs: DANGER HIGH VOLTAGE
For indicating and identifying electrical hazard and high voltage areas
Supplier: Brady™ 89173
View more versions of this product
Safety Services Available
Overview
Logo/Customization
Description
- Warns against entry, machine operation and buried cables
- Made from self-sticking polyester or Aluminum
- Black/red on white
Specifications
11.4W x 5.71cm H (4.5 x 2.25 in.) | |
Black/Red on White | |
1 Ea. | |
Warns against entry, machine operation and buried cables | |
5.71 cm | |
4.5 in. |
DANGER HIGH VOLTAGE | |
Polyester | |
Self-sticking Polyester | |
2.25 in. | |
Admittance Sign | |
11.4 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title