Learn More
Brady™ Air Leak Inspection Tag
Durable Air Leak Inspection Tags inform employees of the inspection history and status of equipment or materials.
Manufacturer:  Brady™ 86562
 View more versions of this product
Catalog No. 19-040-417
Description
Description
  • Write-on polyester laminate (B-837) protects the graphics, wipes clean and withstands water, grease and extreme temperatures.
  • (B-851) Rigid Economy Polyester textured write-on surface allows for authorized signatures, dates and other special instructions.
  • The excellent write-on surface of (B-853) Cardstock is ideal for adding on-the-spot customized notes and other information, recommended for indoor use only.
Specifications
Specifications
ASTM D1593, ASTM D638
Polyester
For marking on-the-spot warning of dangerous conditions
5.75 in.
10 nylon fasteners
-40° to 120°C
3 in.
5.75 in. H x 3 in. W
0.37 in.
Blue on White
Header: Air Leak, Front: 1. Fill Out Work Order/2. Enter Work Order Number On Back Side Of Tag/3. Hang Tag On All Air Leaks, Back: Name/Date/Work Order Number
3 to 5 Years
14.6cm
0.36mm
-40° to 248°F
7.62cm
5.75 in. H x 3 in. W
0.93cm