Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Aisle Marking Tapes
Ultra strong B-726 abrasion-resistant vinyl Aisle Marking Tape makes your facility safer, more visual and more efficient by adding colorful visual indicators to aisles and passageways.
Supplier: Brady™ 58222
Description
- Pressure sensitive rubber based adhesive sticks tightly to virtually any clean, dry surface
- Good chemical, oil and water resistance
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Quick application
Specifications
Acrylic | |
Tape | |
Marking Tape | |
For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification | |
1 Ea. | |
-20°C to +75°C | |
36 yd. | |
2 in. |
Black | |
Vinyl | |
Non-sterile | |
3 Years | |
-4 to 167°F | |
0.004 in. | |
33 m | |
50.8 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction