Brady™ ALLWIK™ Pillows
For absorbing spills and leaking fluids in tight spaces
Manufacturer: Brady™ AW99
 View more versions of this product
Catalog No. 19-000-167
  Check Availability
Add to Cart
    Specifications
    Dimensions (L x W) 25.4 x 25.4cm (10 x 10 in.)
    Product Type ALLWIK Pillow
    For Use With (Application) Overspray Leaks and Drips, Drums - Liquid Storage, Drains
    Quantity 32/Cs.
    Absorbancy 14 gal/bale
    • Absorbs all oil and water-based fluids
  • Description & Specifications
Related Products
Product Content Feedback