Brady™ B-7569 Pre-Printed GHS/CLP Chemical Labels Available on GSA/VA Contract for Federal Government customers only.
Designed with vinyl film that withstands exposure to moisture, chemicals and handling
Manufacturer: Brady™ B302617569CLP3A
 View more versions of this product
Catalog No. 19-140-725
  Check Availability
Add to Cart
Required Accessories : MarkWare™ Software v3.9.1
    Specifications
    For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, and BBP37 printers
    Display Text Legend: L3A Diamond
    For Use With (Application) Chemical labeling
    Quantity 270/Roll
  • Description & Specifications
Related Products
Product Content Feedback