Brady™ BASIC™ Universal Pads

General purpose sorbent to clean up oils, coolants, solvents, and water-based fluids

Manufacturer: Brady™ BPU100

 View more versions of this product

Catalog No. 17-111-144

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Clean up oils, coolants, solvents, and water-based fluids
    Packaging Quantity 100/bale
    Capacity (English) Sorbing 20.5 gal./bale
    Absorbancy Heavy, 20.5 gal./bale
    Color Gray
    Product Type BASIC Universal pad
    Dimensions (L x W) 38.1 x 43.1cm (15 x 17 in.)

    • Sorbents absorb up to 20 times their weight
    • Single-ply meltblown polypropylene
    • Perforated and dimpled
    • Absorbencies: Heavy and light weight
    • Size: 38.1 x 43.1cm (15 x 17 in.)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit