Learn More
Brady™ BBP™11 Slide Label Printers
A low-cost solution for laboratories that are moving toward print labels and bar code label printers, or general industrial companies with low- to medium-volume label printing
Manufacturer:  Brady™ BBP1134L
 View more versions of this product
Catalog No. 22-500-563
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
RoHS, WEEE
Data communications labeling, general and industrial labeling, healthcare identification, laboratory identification, panel identification, wire and cable marking
Bar code label printer, free Brady 360™ 5 year warranty
25.8cm
External universal switching power supply, connectivity: serial, parallel, USB, network (ethernet)
Labelmark™ software, CodeSoft™ software, Windows™ based driver for 3rd party software use
2.49kg
5 Years
Each
6.81 in.
17.3cm
10.16 in.
2MB Flash memory, 8MB SDRAM, SD card slot
300dpi
5.51 lb.
7.95 in.
Slide Label Printer