Learn More
Brady™ BBP™31 Sign and Label Makers
Versatile printer prints signs and labels up to 4 in. (10.2cm) wide
Manufacturer:  Brady™ BBP31360PMWL
 View more versions of this product
Catalog No. 19-211-182
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Blue,Gray
Touch Screen
BBP™30 series thermal transfer tape cartridges and ribbons
AC power, stand alone or PC connectivity via USB
MarkWare™ software, Windows™ based driver for 3rd party software use
6.3kg
Each
RoHS 2002/95/EC, 2005/618/EC MCV, NFPA
Arc flash labeling, barcode labeling, data communications labeling, facility, safety and maintenance identification, laboratory identification, lean/5S labeling, lockout tagout labeling
Brady 360™ preferred plan and MarkWare™ lean software, printer, power cord, USB cable, drivers CD, stylus, cleaning kit, cutter cleaning tool, quick start guide and documentation holder
300dpi
14 lb.
5 Years