Learn More
Brady™ BBP™ Non-Adhesive Tag Stock Supply
Manufacturer:  Brady™ B30C2500551WT
 View more versions of this product
Catalog No. 19-211-273
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
OSHA 1910.145
2 Years
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
-40° to 193°F
2.5 in.
White
Matte
50 ft.
-104° to 379.4°C
Polyester