Brady™ BBP™ Supply Kit

Manufacturer: Brady™ 142062

 View more versions of this product

Catalog No. 19-211-184

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, and BBP37 printers
    Format Tape
    For Use With (Application) Signs, machine warnings, safety warnings, hazardous chemicals (NFPA704M, color, bar, WHIMIS and ANSI formats), emergency egress marking
    Product Type Safety and compliance supply Kit
    Includes One each 4 x 6 in. ANSI header Danger, Warning, Caution and Notice, (2) black and (1) red ribbons, (1) 4 in. photoluminescent tape, (1) 4 x 6 in. RTK NFPA Diamond label with text
    Quantity 1 Pack
    Color Multi-color

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit