Learn More
Brady™ BBP33 Label Printer
Simple, powerful and fast
Manufacturer:  Brady™ BBP33CLMPMWS
 View more versions of this product
Catalog No. 19-140-595
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Printer, power cord, USB cable, drivers CD, stylus, cleaning kit, cutter cleaning tool, quick start guide and documentation holder
Description
Description
Full hassle-free operation with automatic label set-up, no calibration, no sensor adjustment and no label waste.
  • Foolproof drop-in ribbon and cartridge labels – drop-in cartridge, lock and start typing
  • Wide range of material lineup — nearly 500 parts and 39 materials; Product ID, OEM labels and Circuit Board ID; Safety, Facility, Lean5S and Pipemarkers
  • Single color printing
  • Can print both continuous and die cut labels
  • PC connectivity and peripheral printing
  • Resolution: 300 DPI
  • Weight: 5.76kg
  • Print Speed: 7.62cm( 3 in.) second
  • Auto cutter
  • AC power
  • USB, Ethernet and Wireless (optional)
BBB33-C-LMP
  • LabelMark™ 5 software
BBB33-C-MWS
  • MarkWare™ software
BBB33-C-LMP-MWS
  • LabelMark 5 and MarkWare software
Warranty and Services
Brady360™ 5-year warranty — covers repair parts and labor, one print head replacement, a free loaner printer, a stress-free printer setup session and more
Specifications
Specifications
Arc flash labeling, barcode labeling, circuit board labeling, data communications labeling, electrical labeling, facility, safety and maintenance identification, laboratory identification, lean/5S labeling, lockout tagout labeling, panel identification, pipe and valve marking, product identification, security identification, wire and cable identification
AC power, USB, Ethernet, wireless
MarkWare™ software, Labelmark™ software, CodeSoft™ software, Windows™ based driver for 3rd party software use
5.3 kg
Each
Auto cutter, printer, MarkWare™/Labelmark™ software, power cord, USB cable, drivers CD, stylus, cleaning kit, cutter cleaning tool, quick start guide and documentation holder, drop-in cartridge
300dpi
11.7 lb.
5 Years