Learn More
Brady™ Blank Accident Prevention Tags
Blank economy polyester (B-851) accident prevention tags are perfect for marking on-the-spot warnings of dangerous conditions.
Supplier:  Brady™ 76197
 View more versions of this product
Catalog No. 19-102613
Add to Cart
Description
Description
  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Tags include 3/8 in. rolled brass eyelets and come complete with nylon fasteners
  • Withstands moisture, grease and dirt
Specifications
Specifications
ASTM D1593, ASTM D638
Polyester
3 to 5 Years
14.6cm
-40°C to 120°C
Style 2
7.62 cm
0.37 in.
Yellow
For marking on-the-spot warning of dangerous conditions
5.75 in.
0.36 mm
-40°F to 248°F
3 in.
5.75 x 3 in.
0.93 cm
Product Suggestions
Product Suggestions