Brady™ Blank Hazardous Communication Panels

Blank panels to create customized signs

Manufacturer: Brady™ 58502

 View more versions of this product

Catalog No. 19-016239

  Check Availability
Add to Cart

Required Accessories : Quick-Align™ numbers (0 through 4) and symbols (Oxygen, No Water, Radiation, Acid, Corrosive, and Alkali) needed to customize signs sold separately in sizes from 1 to 6 in. . Contact your Fisher Safety Customer Service Representative for ordering information.

    Specifications

    Height (English) 7.5 in.,7.5 in.
    Material Vinyl
    Message NFPA Diamond,NFPA Diamond
    Quantity 5 Pack
    Type Hazardous communication label
    Width (English) 7.5 in.,7.5 in.
    Color Black, Red, Blue, Yellow on White,Black, Red, Blue, Yellow on White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit