Learn More
Brady™ Blank Hazardous Communication Panels
Blank panels to create customized signs
Supplier: Brady™ 58503
Requires : Quick-Align™ numbers (0 through 4) and symbols (Oxygen, No Water, Radiation, Acid, Corrosive, and Alkali) needed to customize signs sold separately in sizes from 1 to 6 in. . Contact your Fisher Safety Customer Service Representative for ordering information.
Description
Make signs to identify hazardous chemical containers and storage areas.
- Place directly on storage tanks, processing tanks and vessels, feed hoppers, pipelines, and other potentially hazardous equipment
- Upper three corners of signal rate health, fire and reactivity on a scale of 0 to 4; lower corner provides specific hazard information
- Available in vinyl with permanent pressure-sensitive adhesive or in rigid fiberglass with predrillled holes for mounting to walls
Compliance
Signals meet NFPA Code 704M and Federal Standard 313.
Specifications
Vinyl | |
10 in. | |
NFPA Diamond | |
Red |
5/Pk. | |
10 in. | |
Hazardous communication label |