Learn More
Brady™ BMP™ 71 PermaSleeve™ Wire Marker Sleeves
Fits snuggly around wire for permanent identification and protection
$337.00 - $741.00
Specifications
Product Type Wire Marker Sleeve
For Use With (Application) Wire and cable marking, Panel identification
Material Polyolefin (B-342)
Certifications/Compliance SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202, RoHS Compliant, UL Recognized; Tested per: ASTM E162 and ASTM E662
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (Metric) Diameter (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Diameter (English) Price Quantity & Availability  
19-107-885
View Documents
Brady™
BM71C125342
1.17 to 2.79 mm 0.046 to 0.110 in.
Each for $337.00
Add to cart
19-107-886
View Documents
Brady™
BM71C187342
1.57 to 3.81 mm 0.062 to 0.150 in.
Each for $360.50
Add to cart
19-107-887
View Documents
Brady™
BM71C250342
2.39 to 5.46 mm 0.094 to 0.215 in.
Each for $444.00
Add to cart
19-107-888
View Documents
Brady™
BM71C375342
3.2 to 8.13 mm 0.0125 to 0.320 in.
Each for $613.50
Add to cart
19-107-889
View Documents
Brady™
BM71C500342
4.75 to 11.43 mm 0.187 to 0.450 in.
Each for $741.00
Add to cart
Description
Description
  • Has 3:1 shrink ratio 
  • Heat shrinkable 
  • Fade resistant
  • Flame retardant
  • Made of B-342 Permasleeve PS polyolefin material
Specifications
Specifications
Wire Marker Sleeve
Wire and cable marking, Panel identification
Polyolefin (B-342)
SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202, RoHS Compliant, UL Recognized; Tested per: ASTM E162 and ASTM E662