Learn More
Brady™ BMP™51/BMP™53 Label Maker Cartridges: Freezerbondz II Polyester Available on GSA/VA Contract for Federal Government customers only.
Ideal identification solutions for biotechnology, agricultural, environmental and forensic research industry
$69.50 - $101.10
Specifications
Adhesive Type Acrylic, Permanent
Color Black, White
Finish White film with white thermal transfer printable topcoat
Life Span 2 Years
View More Specs
Products 11
Catalog Number Mfr. No. Quantity Size (English) Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Size (English) Size (Metric) Price Quantity & Availability  
19-155-142
View Documents
Brady™
M130492
220 labels 0.375 x 0.825 in. 0.95 x 2.1 cm
Each for $90.00
Add to cart
19-155-141
View Documents
Brady™
M82492
240 labels 0.375 x 0.375 in. 0.95 x 0.95 cm
Each for $84.00
Add to cart
19-155-143 Edge
View Documents
Brady™
M131492
180 labels 0.500 x 1 in. 1.27 x 2.54 cm
Each for $96.10
Edge
Add to cart
19-155-144
View Documents
Brady™
M156492
180 labels 1 x 0.9 in. 25.4 x 22.86 mm
Each for $94.10
Add to cart
19-155-148
View Documents
Brady™
M78492
100 Labels 1.9 x 1 in. 4.83 x 2.54 cm
Each for $101.10
Add to cart
19-155-147
View Documents
Brady™
M117492
240 labels 0.5 x 1 in. with 0.4375 in. dia. vial top 1.27 x 2.54 cm
Each for $101.10
Add to cart
19-155-145 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Brady™
M118492
240 labels 0.375 x 1 in. with 0.375 in. dia. vial top 9.53 x 25.4 mm
Each for $98.00
Add to cart
19-155-146 Edge Available on GSA/VA Contract for Federal Government customers only.
View Documents
Brady™
M120492
240 labels 0.5 x 1 in. with 0.375 in. dia. vial top 1.27 x 2.54 cm
Each for $101.10
Edge
Add to cart
19-140-402 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Brady™
MC500492
1 cartridge 0.3 x 0.5 in. 0.762 x 1.27 cm
Each for $97.00
Add to cart
19-000-130
View Documents
Brady™
M125492
180 labels 1.73 x 1 in. 4.4 x 2.54 cm
Each for $69.50
Add to cart
19-000-156
View Documents
Brady™
M95492
360 labels 1.6 x 0.59 in. 4.1 x 1.52 cm
Each for $87.10
Add to cart
Description
Description
Ideal identification solutions for biotechnology, agricultural, environmental and forensic research industry
  • Designed to “match” existing vial, slide, and well plate sizes
  • Holds up under tissue processing protocols and other extreme conditions
  • Readable and allow for easy bar coding and scanning
  • Color: Black on White
  • Material: Freezerbondz™ II Polyester
  • Printer Compatibility: BMP51, BMP53
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Acrylic, Permanent
Black, White
White film with white thermal transfer printable topcoat
2 Years