Brady™ BMP™53 Label Maker Printer
Compact, versatile portable printer with built-in Bluetooth™ technology for wireless printing
Manufacturer: Brady™ BMP53
 View more versions of this product
Catalog No. 19-155-101
  Check Availability
Add to Cart
    Specifications
    Warranty 1 Year(s)
    Display Backlit LCD
    For Use With (Application) Audio or visual labeling, data communications labeling, electrical labeling, facility, safety and maintenance identification, laboratory identification, product identification
    Includes BMP™53 printer, USB cable, MC-1500-595-WT-BK label cartridge, hardcase, quick start guide and AC adapter/battery charger
    Quantity Each
    Resolution 300 dpi
    Weight (English) 2.7 lb.
    Weight (Metric) 1.22kg
    Connector Types USB, Bluetooth and optional Ethernet, Wi-Fi
  • Description & Specifications
Related Products
Product Content Feedback