Learn More
Brady™ BMP™53 Label Maker Printer
Compact, versatile portable printer with built-in Bluetooth™ technology for wireless printing
Manufacturer:  Brady™ BMP53KITLS
 View more versions of this product
Catalog No. 19-018-018
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
USB, Bluetooth and optional Ethernet, WiFi
Audio or visual labeling, data communications labeling, electrical labeling, facility, safety and maintenance identification, laboratory identification, product identification
300dpi
1.22kg
1 Kit
Backlit LCD
BMP™53 printer, USB cable, hardcase, quick start guide, AC adapter/battery charger, Labelmark™ standard software and M-156-492, M-81-48, M-120-492, M-103-498, MC-1500-595-WT-BK label cartridges
2.7 lb.
1 Year(s)