Learn More
Brady™ BMP53 Label Maker kit
BMP53 Label Maker kit with built-in Bluetooth™ technology for wireless printing.
Supplier:  Brady™ BMP53KITACBP
Catalog No. 19-000-641
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • The BMP™53 Printer is ideal for electrical, catacomb, facility/safety, product and panel manufacturing, and laboratory labeling applications
  • Features a 300 dpi print head and can print professional, durable labels up to 1.5 in. wide
  • Prints on continuous and die-cut materials at a speed of 1 inch per second and uses the same label cartridges as the BMP™51 printer
  • Offers three power options: AC Power (included), rechargeable Lithium Ion Battery (included), or 8 AA's (not included)
Warranty and Services
2 Year Limited
Specifications
Specifications
152.4 mm
6 x 6 x 3.6 in.
Cable and wire marking, Facility ID, Healthcare ID, Laboratory ID, Lean/5S, maintenance labeling, Panel manufacturing, Product ID, Regulatory Compliance labeling, Safety labeling, Visual workplace labeling
6 in. H x 6 in. W x 3.6 in. D
Non-sterile
Product Suggestions
Product Suggestions