Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Bradylink™ Solid Color Warning Posts

Provides sturdy support for barricading chain and tapes

Manufacturer:  Brady™ 92120

 View more versions of this product

Catalog No. 19-104-360


Add to cart

Description

Description

  • Posts are non-conductive so can be safely used in electrically hazardous areas
  • Hollow base can be filled with sand for more stable application
  • Large C-hook holds chain in place on large posts 
  • Can be used open or closed for greater security
Specifications

Specifications

6/Pk.
3 in.
White
7.62cm
104.1cm
Polyethylene base and top, Polystyrene post
Safety
16 in.
40.6cm
Warning Post
41 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit