Learn More
Brady™ Breaker Lockout Sampler Pouch with Safety Padlocks and Tags
Breaker Lockout Sampler Pouch with Safety Padlocks and Tags is equipped with the tools you need to isolate mechanical and electrical control points.
Supplier: Brady™ 99296
Safety Services Available
Overview
LOTO Assessments
Description
- The Breaker Lockout Sampler Pouch contains one of each of our most popular circuit breaker lockouts
- Comes in a highly visible, easy to transport red pouch
- Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
- Brady™ is a world leader in lockout/tagout products and services offering industry expertise and a broad range of durable, easy-to-use lockout devices
Specifications
Breaker Lockout Sampler Pouch with Safety Padlocks and Tags | |
OSHA 1910.147 | |
2 in. | |
(1) 120V Snap-On Breaker Lockout, (1) 120/277V Clamp-On Breaker Lockout, (1) 480/600V Clamp-On Breaker Lockout, (1) Universal Multi-Pole Breaker Lockout, (1) Oversized Breaker Lockout, (1) Cleat for 120/277V Clamp-On Lockout, (1) Cleat for 480/600V Clamp-On Lockout, (1) Lockout Belt Pouch, 2-Brady Safety Padlocks and 2-Lockout Tags and nylon cable ties |