Learn More
Brady™ BSP™ 31 Label Attachment System and BBP™11 Tissue Cassette Starter Kit with Brady™ 360 Basic Program
Permanently attaches labels to tissue cassettes, resulting in more accurate specimen tracking and sample management
Manufacturer:  Brady™ BSP31BBP11KIT1
Catalog No. 22-501-731
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Compatible with 35 and 45° angle Fisherbrand™, Leica™, Simport™, and Sakura™ Tissue-Tek™ tissue cassettes
  • Preprocess identification for tissue samples allows traceability throughout laboratory processes and storage
  • Reduce the chance of identification errors and reduce the liability
  • Ideal as a backup system for high volume laboratories
Specifications
Specifications
Label attachment and tissue cassette starter Kit
Healthcare identification, laboratory identification, tissue cassette labeling, ideal as a backup system for high volume laboratories
5 Years
5.89 kg
6.61 x 10.34 x 6.45 in. (16.78 x 26.26 x 16.38 cm)
Brady 360 basic program, BBP™11 replacement program, 1-R6413 ribbon and 1-1.0 in. core tissue label cassette
13 lb.
1 Ea.