Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “Caution: Chemical Goggles Required…” Laboratory Signs Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  Brady™ 69041

 View more versions of this product

Catalog No. 19-102-425


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

OSHA 1910.145, ASTM D 1593, ASTM D 638, ASTM D 790, ASTM D 646, ASTM D 635
Industrial, utility, commercial and institutional environment
Premium Fiberglass
Each
-40 to 193°F
9000psi
Eye protection sign
Black on Yellow
10 in.
Caution Chemical Goggles Required In This Area
Surface Mount
-40 to 90°C
0.1 in.
14 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit