Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “Caution: Eye and Glove Protection...” Signs Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  Brady™ 84302

 View more versions of this product

Catalog No. 19-035671


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ANSI Z535, OSHA 1910.145, PSTC-33,
Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
Polyester
Each
-40 to 230°F
0.006 to 0.01 in.
14 in.
Black on Yellow
10 in.
Caution Eye And Glove Protection Must Be Worn When Handling Chemicals
Surface Mount
-40 to 110°C
Chemical and hazardous materials sign
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit