Brady™ “Caution: Personal Protective Clothing...” Signs Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Brady™ 84315

 View more versions of this product

Catalog No. 19-035681

  Check Availability
Add to Cart

    Specifications

    Height (English) 7 in.,7 in.
    Material Polyester
    Certifications/Compliance ANSI Z535, OSHA 1910.145, PSTC-33,,ANSI Z535, OSHA 1910.145, PSTC-33,
    Message Caution Personal Protective Clothing Is To Be Worn At All Times When Handling Chemicals
    Quantity Each
    Style Surface Mount
    Temperature Range (Metric) -40 to 110°C
    Thickness 0.006 to 0.01 in.
    Type Chemical and hazardous materials sign
    Width (English) 10 in.
    For Use With (Application) Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
    Temperature Range (English) -40 to 230°F
    Color Black on Yellow,Black on Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit