Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “Caution: Stand Clear of Door” Signs Available on GSA/VA Contract for Federal Government customers only.

Door sign with message for workplace

Manufacturer:  Brady™ 84711

 View more versions of this product

Catalog No. 19-035-858


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ANSI Z535, OSHA 1910.145
Used for safety signs, identification of fire alarms, extinguishers and emergency exits
25.4cm
Caution Stand Clear Of Door
Surface Mount
-40 to 110°C
Door sign
35.56cm
Black on Yellow
10 in.
B-302 Self Sticking Polyester
Each
-40 to 230°F
0.006 to 0.01 in.
14 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit