Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Caution and Warning Tapes Available on GSA/VA Contract for Federal Government customers only.

Various tape designs in choice of two materials

Manufacturer:  Brady™ 76316

 View more versions of this product

Catalog No. 19-807-242


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

54 ft. x 2 in. (16.45m x 5.08cm)
Black,White
Warning Stripe and Check Tape
For marking physical hazards
-40° to 160°F
0.15mm
5.08cm
OSHA 1910.144
Vinyl
54 ft.
16.45m
- 40° to 71°C
2 in.
Rubber based pressure sensitive

Safety and Handling

ShelfLife : 1 to 2 Years

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit