Brady™ Caution and Warning Tapes

Various tape designs in choice of two materials

Manufacturer: Brady™ 76432

 View more versions of this product

Catalog No. 19-807-244

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Length (English) 5 yd.
    For Use With (Application) For marking physical hazards
    Shelf Life 1 to 2 Years
    Length (Metric) 4.57m
    Width (English) 2 in.
    Width (Metric) 5.08cm
    Adhesive Rubber based pressure sensitive
    Color Solid White, Glow-in-the-Dark
    Material Glow-in-the-Dark vinyl
    Product Type Warning Stripe and Check Tape
    Dimensions (L x W) 5 yd. x 2 in. (4.57m x 5.08cm)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit