Brady™ Chemical and Hazardous Materials Signs: DANGER - EXPLOSIVES Available on GSA/VA Contract for Federal Government customers only.

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

Manufacturer: Brady™ 85167

 View more versions of this product

Catalog No. 19-035-935

  Check Availability
Add to Cart

    Specifications

    Style Surface Mount
    Temperature Range (English) -40 to 230°F
    Temperature Range (Metric) -40 to 110°C
    Color Red, Black on White,Red, Black on White
    Height (English) 7 in.
    Height (Metric) 17.78cm
    Material Polyester
    Certifications/Compliance ANSI Z535, OSHA 1910.145
    Message Explosive
    Quantity Each
    Thickness 0.006 to 0.01 in.
    Type Chemical and hazardous materials sign
    Width (English) 10 in.
    Width (Metric) 25.40cm
    For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit