Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Chemical and Hazardous Materials Signs: DANGER - FLAMMABLE LIQUIDS
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
$26.00 - $96.00
Specifications
Certifications/Compliance | ANSI Z535, OSHA 1910.145 |
---|---|
Quantity | 1 Ea. |
Mounting Type | Surface Mount |
For Use With (Application) | Designed for use in industrial, utility, commercial and institutional environments |
Min. Temperature (English) | -40°F |
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
19-016865
|
Brady™
72238 |
Fiberglass | 10 in. | 14 in. |
Each for $96.00
|
|
|||
19-035711
|
Brady™
25665 |
Plastic | 10 in. | 14 in. |
Each for $26.00
|
|
|||
19-035961
|
Brady™
85182 |
Polyester | 10 in. | 14 in. |
Each for $66.00
|
|
|||
19-036-512
|
Brady™
85181 |
Polyester | 7 in. | 10 in. |
Each for $31.75
|
|
|||
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in various widths and heights
- Black/red on white
Specifications
ANSI Z535, OSHA 1910.145 | |
Surface Mount | |
-40°F | |
Hazardous Materials Sign |
1 Ea. | |
Designed for use in industrial, utility, commercial and institutional environments | |
Flammable Liquid | |
Red on White |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title