Brady™ Chemical and Hazardous Materials Signs: Diesel Fuel

For identifying plant's chemical storage and hazardous materials area

Manufacturer: Brady™ 70266

 View more versions of this product

Catalog No. 19-016610

  Check Availability
Add to Cart

    Specifications

    Grade Industrial
    Height (English) 10 in.,10 in.
    Material Premium Fiberglass
    Certifications/Compliance ISO 9001
    Message Danger Diesel Fuel
    Quantity Each
    Style Surface Mount
    Temperature Range (English) -40 to 193°F,-40 to 193°F
    Temperature Range (Metric) -40 to 90°C,-40 to 90°C
    Tensile Strength 9000psi
    Thickness 0.1 in.,0.1 in.
    Color Black, Red on White,Black, Red on White
    Type Chemical and hazardous materials sign
    Width (English) 14 in.,14 in.
    For Use With (Application) Industrial, utility, commercial and institutional environment

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit