Learn More
Brady™ CodeXML™ M3 Bluetooth Barcode Reader
Synchronizes up to three scanners to one computer
Manufacturer:  Brady™ 102890
Catalog No. 11-878-514
Add to cart
Description
Description
  • Simple to set up
  • Has a range of 300 feet
  • Bluetooth operation auto-enabled by scanning a barcode printed on the modem (must purchase separately)
  • 6.8 in. LCD Monitor
  • 110VAC Only
Compliance
RoHS Directive
Specifications
Specifications
CodeXML M3 Bluetooth Barcode Reader
Bluetooth™, USB
For Bluetooth™ enabled CR2, CR2500, CR3 and CR3500 barcode scanners, Wide linear and 2D matrix barcodes
RoHS
LCD