Brady™ CR3500 Barcode Scanners
Decodes bar codes faster with familiar cell phone style keypad
Manufacturer: Brady™ CR3512GBH1R0C0
 View more versions of this product
Catalog No. 11-878-513
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Connection Type Bluetooth™, USB, RS232 or PS/2
    Data Memory 10MB
    Includes CR3500 Barcode Scanner with built-in Bluetooth™ radio, 1950 mAH Li-Ion rechargeable BH1 battery handle and 6ft straight USB cable
    Product Type CR3500 Barcode Scanner
    Lightweight, ergonomic design for unequaled performance on difficult reading surfaces, including circuit boards, IV bags, patient-wristbands, driver licenses and other shiny or curved surfaces.
    • Features MegaPixel digital imaging, dual-field optics and glare reducing technology
    • Features high-speed omni-directional decoding
    • Decodes high density matrix codes and larger low density linear codes
    • Comes with cordless and cabled interfaces
    • Alpha-numeric keypad and display screen for manual data entry
    • Equally efficient in batch, cabled or wireless modes
    • Features 10MB of non-volatile memory for saving batch data
    • Good read feedback with vibration, audible tones and LED indicators
    • Firmware is field upgradeable
    • Data rates are high
    • Reads and decodes all symbologies
    • Data ouput via Bluetooth, USB, Serial (RS-232) or PS2 options
    • JavaScript Application Development Suite allows program development to allow advanced data manipulation
  • Description & Specifications
Related Products
Product Content Feedback