Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ “Danger: Confined Space -- Hazardous Atmosphere” Signs
For identifying areas that require authorized or permit entry only
Supplier: Brady™ 84567
View more versions of this product
Safety Services Available
Overview
Logo/Customization
Description
- Bright colors and bold text provide high visibility to critical messages
- Legend: CONFINED SPACE HAZARDOUS ATMOSPHERE CHECK OXYGEN LEVEL BEFORE AND DURING ENTRY
- Header: DANGER
- Black/red on white
- Designed to withstand harsh industrial environments
- Available in different sizes and materials
B-401
- Plastic — high-impact polystyrene
- Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
Specifications
ANSI Z535, OSHA 1910.145 | |
1 Ea. | |
Surface Mount | |
7 in. | |
230°F | |
0.006 to 0.01 in. | |
25.4 cm | |
-40°F to 230°F | |
Confined Space Hazardous Atmosphere Check Oxygen Level Before And During Entry | |
Red on White |
Polyester | |
Self-sticking Polyester (B-302) | |
Designed for use in industrial, utility, commercial and institutional environments | |
17.78 cm | |
-40°F | |
10 in. | |
-40°C to 110°C | |
25.4 x 17.78cm (10 x 7 in.) | |
Admittance Sign |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction