Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “Danger: Do Not Enter -- Emergency Exit Only” Signs Available on GSA/VA Contract for Federal Government customers only.

For identifying areas that require authorized or permit entry only

Manufacturer:  Brady™ 22084

 View more versions of this product

Catalog No. 19-035588


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ANSI Z535, OSHA 1910.145
For use in light duty industrial, utility, commercial and institutional environments
25.4cm
Danger Do Not Enter Emergency Exit Only
Surface Mount
-40 to 80°C
Admittance sign
35.56cm
Black, Red on White
10 in.
B-401 Plastic
Each
-40 to 175°F
0.06 in.
14 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit