Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ “Danger: Hardhats, Ear And Eye Protection Required...” Signs Available on GSA/VA Contract for Federal Government customers only.

Provides strong and noticeable reminder for your employees to protect themselves and maintain safe work atmosphere

Manufacturer:  Brady™ 75979

 View more versions of this product

Catalog No. 19-036-300


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ASTM D 1593, D 638, D 790, D 646, D 635, ANSI Z535, OSHA 1910.145
Industrial, utility, commercial and institutional environments for both indoor and outdoor use
17.7cm
Danger Hardhats, Ear And Eye Protection Required Beyond This Point
Surface Mount
-40 to 90°C
0.1 in.
10 in.
Black, Red on White
7 in.
Premium Fiberglass
Each
-40 to 193°F
9000psi
Protective wear sign
25.4cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit